Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

D141208

Sigma-Aldrich

2-Chloro-N,N-dimethylethylamine hydrochloride

99%

Synonym(s):

1-Chloro-2-dimethylaminoethane hydrochloride, 2-(Dimethylamino)ethyl chloride hydrochloride, N-(2-Chloroethyl)dimethylamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCH2CH2Cl · HCl
CAS Number:
Molecular Weight:
144.04
Beilstein:
3591234
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

crystals

mp

201-204 °C (lit.)

SMILES string

Cl.CN(C)CCCl

InChI

1S/C4H10ClN.ClH/c1-6(2)4-3-5;/h3-4H2,1-2H3;1H

InChI key

LQLJZSJKRYTKTP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Muta. 2 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service