Skip to Content
MilliporeSigma

8.02448

N-(2-Chloroethyl)-N,N-dimethylammonium chloride

for synthesis

Synonym(s):

N-(2-Chloroethyl)-N,N-dimethylammonium chloride, Dimethylaminoethyl chloride hydrochloride, N,N-Dimethyl-N-2-chloroethylammonium chloride

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Linear Formula:
ClCH2CH2(CH3)2N · HCl
CAS Number:
Molecular Weight:
144.04
UNSPSC Code:
12352107
EC Index Number:
224-970-1
NACRES:
NA.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

N-(2-Chloroethyl)-N,N-dimethylammonium chloride, for synthesis

SMILES string

[Cl-].ClCC[N+H](C)C

InChI

1S/C4H10ClN.ClH/c1-6(2)4-3-5;/h3-4H2,1-2H3;1H

InChI key

LQLJZSJKRYTKTP-UHFFFAOYSA-N

form

powder

autoignition temp.

220 °C

potency

238 mg/kg LD50, oral (Rat)
290 mg/kg LD50, skin (Rabbit)

mp

205-208 °C

solubility

2000 g/L

storage temp.

2-30°C

Quality Level

Analysis Note

Assay (argentometric): ≥ 98.0 %
Identity (IR): passes test

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Muta. 2 - STOT RE 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service