Key Documents
CLS9985150
Pyrex® watch glasses
O.D. 150 mm
Synonym(s):
Corning® 9985
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Pricing and availability is not currently available.
manufacturer/tradename
Corning 9985-150
O.D.
150 mm
Related Categories
General description
Heavy-wall, borosilicate glass with fire-polished edge provides durability. May be slightly out-of-round due to O.D. variability.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)