Documents
CLS998575
Pyrex® watch glasses
O.D. 75 mm
Synonym(s):
Corning® 9985
About This Item
UNSPSC Code:
41121800
manufacturer/tradename
Corning 9985-75
O.D.
75 mm
General description
Heavy-wall, borosilicate glass with fire-polished edge provides durability. May be slightly out-of-round due to O.D. variability.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.