Key Documents
CLS70781CN
Pyrex® serological pipettes, disposable, sterile, plugged
volume 1.0 mL, accuracy: 0.01 mL
Synonym(s):
Corning® 7078
About This Item
UNSPSC Code:
41121500
NACRES:
NB.04
material
borosilicate glass
description
plugged
sterility
sterile
manufacturer/tradename
Corning 7078-1CN
volume measuring range
1.0 mL, accuracy: 0.01 mL
capacity
1 mL
General description
These sterile disposable Pyrex serological pipettes are designed for the larger-volume user. They are calibrated "to deliver" (TD) with blow-out and are packed to permit easy access to several pipettes at a time without contaminating the entire case. Similar to No. 7077, except multi-packed in plastic bags.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.