Key Documents
BAF441500005
Scienceware® engraving tool, battery powered
L 17.8 cm (7 in.)
About This Item
UNSPSC Code:
41100000
NACRES:
NB.35
packaging
pkg of 1 ea
manufacturer/tradename
Bel-Art F44150-0005
L
17.8 cm (7 in.)
General description
Engrave test tubes, bottles, racks, keys, equipment, tools, electronics and more. Simply hold the Engraving Tool like a pen, press the power button, apply light pressure, and ˜write′ to permanently mark most materials including glass, plastic, wood, metal, bone, and stone.
  • Portable, cordless battery power; operates on two ′AA′ batteries (not included)
  • Use indoors or out
  • Engraving is permanent; Marks will not fade if exposed to chemicals or sunlight
  • Extra engraving tip included
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP