Key Documents
CLS5400125
Pyrex® iodine determination flask, with Pyrex® ST stopper
capacity 125 mL
Synonym(s):
flask erlenmeyer, flasks erlenmeyer, glass flasks, Corning® 5400
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Pricing and availability is not currently available.
material
borosilicate glass
wide-mouth
Joint
joint ST/NS stopper22
sterility
non-sterile
feature
closure type ST stopper
packaging
case of 12
manufacturer/tradename
Corning 5400-125
capacity
125 mL
General description
Flasks are blown in specillay designed molds, thereby insuring uniformity and proper contour. The stopper projects above the liquid seal trough to facilitate removal.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)