Documents
CLS53404L
Pyrex® heavy wall graduated filtering flask, with tubulation
12.5 mm O.D. hose-connection, capacity 4,000 mL
Synonym(s):
conical flasks, filtering flasks, flasks erlenmeyer, Corning® 5340
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass
flat bottom flask
narrow-mouth
Joint
joint stopper12
sterility
non-sterile
feature
closure type Rubber stopper
packaging
pack of 1
case of 6
manufacturer/tradename
Corning 5340-4L
volume measuring range
1,500-4,000 mL
capacity
4,000 mL
General description
These flasks have sidearm tabulations and are blown in special molds designed to give maximum mechanical strength. For convenience, the flasks are graduated to show approximate capacity. The neck finish affords an excellent fit for rubber stoppers.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.