Documents
CLS53204L
Pyrex® heavy-wall graduated filtering flask
capacity 4,000 mL
Synonym(s):
conical flasks, filter flasks, filtering flask, filtering flasks, flask filter, Corning® 5320
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass
flat bottom flask
narrow-mouth
Joint
joint stopper12
description
without hose connection
sterility
non-sterile
feature
closure type Rubber stopper
packaging
pack of 1
case of 6
manufacturer/tradename
Corning 5320-4L
volume measuring range
1,500-4,000 mL
capacity
4,000 mL
General description
Blown in special molds to give maximum strength. Graduated to show approximate capacity.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.