Skip to Content
MilliporeSigma
All Photos(1)

Documents

CLS44444L

Pyrex® baffled shaker flasks

flask capacity 4,000 mL

Synonym(s):

conical flasks, Corning® 4444

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121804
NACRES:
NB.43

material

Delong
borosilicate glass

sterility

non-sterile

feature

cap: no
closure type Closure cap

packaging

case of 1

manufacturer/tradename

Corning 4444-4L

flask capacity

4,000 mL

neck O.D.

(38 mm)
38 mm

Looking for similar products? Visit Product Comparison Guide

General description

  • Pyrex® baffled shaker flasks are made of optically clear borosilicate glass.
  • Delong-style Erlenmeyer flasks feature three baffle indents to provide greater agitation of solutions to improve oxygen or gas transfer when used with rotary or reciprocating shakers.
  • Heavy, fire-polished, long necks reduce splashing and accept two-position PP closures.
  • Uniform wall thickness ensures an optimal balance between mechanical strength and thermal shock resistance.
  • Includes a marking spot for convenience.

Legal Information

Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service