Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS2679525

Corning® PP closures for cell culture tubes and flasks

Synonym(s):

cell culture flask caps, cell culture tube caps

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

material

polypropylene

description

fits neck O.D. 25 mm

sterility

non-sterile

feature

reusable

packaging

case of 100
pack of 50

manufacturer/tradename

Corning 26795-25

color

natural

Looking for similar products? Visit Product Comparison Guide

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service