Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CDS003904

Sigma-Aldrich

Boc-Lys(Boc)-OH

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H30N2O6
Molecular Weight:
346.42
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

SMILES string

CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(O)=O

InChI

1S/C16H30N2O6/c1-15(2,3)23-13(21)17-10-8-7-9-11(12(19)20)18-14(22)24-16(4,5)6/h11H,7-10H2,1-6H3,(H,17,21)(H,18,22)(H,19,20)/t11-/m0/s1

InChI key

FBVSXKMMQOZUNU-NSHDSACASA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yang Lan et al.
ACS applied materials & interfaces, 12(28), 31904-31921 (2020-06-20)
Immunochemotherapy is viewed as a promising approach for cancer therapy via combination treatment with immune-modulating drugs and chemotherapeutic drugs. A novel dual-functional immunostimulatory polymeric prodrug carrier PEG2k-Fmoc-1-MT was developed for simultaneously delivering 1-methyl tryptophan (1-MT) of an indoleamine 2,3-dioxygenase (IDO)
Haofang Zhu et al.
ACS applied materials & interfaces, 12(4), 4241-4253 (2019-12-21)
Due to the inconvenience of application, risk of extra damage to fragile soft tissues, and the high incidence of late-stage complications, significant research endeavors have been focused on developing safe and effective bioadhesives to replace or assist the traditional suture
Jiban J Panda et al.
Journal of nanobiotechnology, 11, 18-18 (2013-06-27)
Gene therapy is most effective when delivery is both efficient and safe. However, it has often proven difficult to find a balance between efficiency and safety in case of viral or polymeric vectors for gene therapy. Peptide based delivery systems
Yujia Zhang et al.
Journal of materials chemistry. B, 8(33), 7428-7437 (2020-07-15)
Infectious diseases induced by pathogenic bacteria are the major causes for the failure of medical implants. Meanwhile, the drug-resistance is steadily developed because of the large and even inappropriate use of antibiotics. Therefore, the development of antibacterial coating with non-antibiotic-based
Zhan Huang et al.
Biomaterials, 61, 216-228 (2015-05-26)
Tissue regeneration and development involves highly synchronized signals both between cells and with the extracellular environment. Biomaterials can be tuned to mimic specific biological signals and control cell response(s). As a result, these materials can be used as tools to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service