MilliporeSigma
Unsupported Mg-Alkene Bonding.
Chemistry (Weinheim an der Bergstrasse, Germany) (2020-11-17)
Katharina Thum, Alexander Friedrich, Jürgen Pahl, Holger Elsen, Jens Langer, Sjoerd Harder
ABSTRACT
The first intermolecular early main group metal-alkene complexes were isolated. This was enabled by using highly Lewis acidic Mg centers in the Lewis base-free cations (Me BDI)Mg+ and (tBu BDI)Mg+ with B(C6 F5 )4 - counterions (Me BDI=CH[C(CH3 )N(DIPP)]2 , tBu BDI=CH[C(tBu)N(DIPP)]2 , DIPP=2,6-diisopropylphenyl). Coordination complexes with various mono- and bis-alkene ligands, typically used in transition metal chemistry, were structurally characterized for 1,3-divinyltetramethyldisiloxane, 1,5-cyclooctadiene, cyclooctene, 1,3,5-cycloheptatriene, 2,3-dimethylbuta-1,3-diene, and 2-ethyl-1-butene. In all cases, asymmetric Mg-alkene bonding with a short and a long Mg-C bond is observed. This asymmetry is most extreme for Mg-(H2 C=CEt2 ) bonding. In bromobenzene solution, the Mg-alkene complexes are either dissociated or in a dissociation equilibrium. A DFT study and AIM analysis showed that the C=C bonds hardly change on coordination and there is very little alkene→Mg electron transfer. The Mg-alkene bonds are mainly electrostatic and should be described as Mg2+ ion-induced dipole interactions.
Sigma-Aldrich
2-Ethyl-1-butene, 95%
Sigma-Aldrich
Cycloheptatriene, 95%
Sigma-Aldrich
1,3-Divinyltetramethyldisiloxane, 97%