Skip to Content
MilliporeSigma
All Photos(2)

Documents

C94602

Sigma-Aldrich

2-Pyridinecarbonitrile

99%

Synonym(s):

2-Cyanopyridine, Picolinonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4N2
CAS Number:
Molecular Weight:
104.11
Beilstein:
107710
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

refractive index

n20/D 1.529 (lit.)

bp

212-215 °C (lit.)

mp

24-27 °C (lit.)

density

1.081 g/mL at 25 °C (lit.)

SMILES string

N#Cc1ccccn1

InChI

1S/C6H4N2/c7-5-6-3-1-2-4-8-6/h1-4H

InChI key

FFNVQNRYTPFDDP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Juan Amaro-Gahete et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 25(27), 6823-6830 (2019-03-19)
Graphene-based materials exhibit outstanding physical properties and so are potentially applicable in a great variety of fields. Unlike their corresponding oxides, graphite and graphene are not prone to functionalization. Diels-Alder reactions are among the scarce reactions that they can occur
Masazumi Tamura et al.
Nature communications, 6, 8580-8580 (2015-10-06)
Multidentate materials formed by simply mixing heterogeneous and homogeneous components are promising for construction of versatile active sites on the surface of heterogeneous compounds, however, to the best of our knowledge, there are no reports on such materials. Self-assembly of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service