516333
4-Acetylbenzaldehyde
97%
Synonym(s):
4-Formylacetophenone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3COC6H4CHO
CAS Number:
Molecular Weight:
148.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
mp
33-36 °C (lit.)
functional group
aldehyde
ketone
SMILES string
[H]C(=O)c1ccc(cc1)C(C)=O
InChI
1S/C9H8O2/c1-7(11)9-4-2-8(6-10)3-5-9/h2-6H,1H3
InChI key
KTFKRVMXIVSARW-UHFFFAOYSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Low-Pressure Hydrogenation of Arenecarboxylic Acids to Aryl Aldehydes.
Goo?en LJ, et al.
Advanced Synthesis & Catalysis, 352(13), 2166-2170 (2010)
Application of the combination of sodium bisulfite as a protective reagent and solid supports in the selective reduction of 4-acetylbenzaldehyde with diborane.
Chihara T, et al.
Chemistry Letters (Jpn), 11, 1657-1660 (1981)
Selective reduction of less reactive carbonyl groups in the presence of diborane and sodium bisulfite on silica gel.
Chihara T, et al.
Canadian Journal of Chemistry, 68(5), 720-724 (1990)
Bis [(tetraphenylporphyrinato) zinc]-calix [4] pyrrole. Synthesis and receptor properties.
Mamardashvili NZ, et al.
Russ. J. Org. Chem., 50(4), 559-566 (2014)
NaBH4/NaNO3/H2O: A Convenient System for Selective Reduction of Aldehydes VS. Ketones to their Corresponding Alcohols.
Ghaderi S and Setamdideh D.
Orient. J. Chem., 30(4), 1913-1917 (2014)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service