Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C102652

Sigma-Aldrich

2-Cyclohexen-1-ol

95%

Synonym(s):

(±)-2-Cyclohexen-1-ol, 1-Cyclohexen-3-ol, 1-Hydroxy-2-cyclohexene, 2-Cyclohexenyl alcohol, 3-Hydroxy-1-cyclohexene, 3-Hydroxycyclohexene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H9OH
CAS Number:
Molecular Weight:
98.14
Beilstein:
1446187
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.487 (lit.)

bp

164-166 °C (lit.)

density

1 g/mL at 25 °C (lit.)

SMILES string

OC1CCCC=C1

InChI

1S/C6H10O/c7-6-4-2-1-3-5-6/h2,4,6-7H,1,3,5H2

InChI key

PQANGXXSEABURG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

136.4 °F - closed cup

Flash Point(C)

58 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kwon Hee Bok et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 23(13), 3117-3125 (2016-12-22)
A new mononuclear nickel(II) complex, [Ni
Katarzyna Rydel-Ciszek et al.
Chemicke zvesti, 71(11), 2085-2093 (2017-11-07)
Manganese(II) complex [(Bn-tpen)Mn
Yun Wang et al.
Materials (Basel, Switzerland), 12(20) (2019-10-12)
Catalyzed organic solvent-free (ep)oxidation were achieved using H3PM12O40 (M = Mo or W) complexes ionically grafted on APTES-functionalized nano-silica beads obtained from straightforward method (APTES = aminopropyltriethoxysilane). Those catalysts have been extensively analyzed through morphological studies (Dynamic Light Scattering (DLS)
Baocheng Zhou et al.
Molecules (Basel, Switzerland), 20(5), 8429-8439 (2015-05-15)
Iron (III) phthalocyanine complexes were employed for the first time as a mild and efficient Lewis acid catalyst in the selective oxidation of cyclohexene to cyclohexane-1,2-diol. It was found that the catalyst FePcOTf shown excellent conversion and moderate selectivity relative

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service