Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

528692

Sigma-Aldrich

2-(6-Bromohexyloxy)tetrahydro-2H-pyran

97%

Synonym(s):

1-Bromo-6-tetrahydropyranyloxyhexane, 6-Tetrahydropyraryloxy-1-bromohexane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H21BrO2
CAS Number:
Molecular Weight:
265.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

contains

1% potassium carbonate as stabilizer

refractive index

n20/D 1.478 (lit.)

bp

125 °C/0.1 mmHg (lit.)

density

1.209 g/mL at 25 °C (lit.)

functional group

bromo
ether

storage temp.

2-8°C

SMILES string

BrCCCCCCOC1CCCCO1

InChI

1S/C11H21BrO2/c12-8-4-1-2-5-9-13-11-7-3-6-10-14-11/h11H,1-10H2

InChI key

CWSSIUJITPYGLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-(6-Bromohexyloxy)tetrahydro-2H-pyran can be synthesized by reacting 6-bromohexanol and pyridinium p-toluenesulfonate in anhydrous dichloromethane.

Application

2-(6-Bromohexyloxy)tetrahydro-2H-pyran may be used as a reactant in the synthesis of diethyl 6-(tetrahydropyran-2-yloxy)hexylphosphonate and 13-(tetrahydo-2H-pyran-2-yloxy)-6-tridecyn-1-ol.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and polymerization kinetics of acrylamide phosphonic acids and esters as new dentine adhesives.
Besse V, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 51(1), 149-157 (2013)
Ashot Khrimian et al.
Journal of agricultural and food chemistry, 56(7), 2452-2456 (2008-03-13)
The browntail moth, Euproctis chrysorrhea (L.), is native to Eurasia, where periodic outbreaks result in defoliation of forest, shade, and ornamental trees. In addition to the damage caused by defoliation, human contact with larval urticating hairs often results in severe

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service