C102008
1,2-Cyclohexanedione dioxime
96%
Synonym(s):
Nioxime
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6H8(=NOH)2
CAS Number:
Molecular Weight:
142.16
Beilstein:
1909047
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
96%
form
powder
mp
185-188 °C (lit.)
SMILES string
O\N=C1CCCCC/1=N\O
InChI
1S/C6H10N2O2/c9-7-5-3-1-2-4-6(5)8-10/h9-10H,1-4H2/b7-5+,8-6+
InChI key
CUNNCKOPAWXYDX-KQQUZDAGSA-N
Application
Forms complexes with Co, Fe and Ni.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Min Liu et al.
Dalton transactions (Cambridge, England : 2003), 46(42), 14509-14518 (2017-11-01)
In this study, a new
Min Liu et al.
Nuclear medicine and biology, 43(11), 732-741 (2016-10-25)
In this study, novel Complexes [ Both azide coligand and boronate caps had significant impact on the heart uptake and myocardial retention of complexes [
Yumin Zheng et al.
Nuclear medicine and biology, 41(10), 813-824 (2014-08-30)
(99m)Tc-Teboroxime ([(99m)TcCl(CDO)(CDOH)2BMe]) is a member of the BATO (boronic acid adducts of technetium dioximes) class of (99m)Tc(III) complexes. This study sought to explore the impact of co-ligands on solution stability, heart uptake and myocardial retention of [(99m)Tc(L)(CDO)(CDOH)2BMe] ((99m)Tc-Teboroxime: L=Cl; (99m)Tc-Teboroxime(F):
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service