Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BR72376

BRAND® bucket, PP

volume 12 L, H 300 mm

Synonym(s):

Buckets, Containers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122800
NACRES:
NB.51

packaging

pack of 1 ea

manufacturer/tradename

BRAND 72376

H

300 mm

volume

12 L
12000 mL

Looking for similar products? Visit Product Comparison Guide

General description

Without lid and with spout and reinforced rim and handle.

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service