Key Documents
BR65238
BRAND® desiccator ′Novus′
nominal diameter × O.D. × H 200 mm × 270 mm × 368 mm, top socket joint: 24/29
About This Item
UNSPSC Code:
41104510
NACRES:
NB.43
Joint
socket joint: top24/29
feature
lid with socket and stopcock
manufacturer/tradename
BRAND 65238
nominal diameter × O.D. × H
200 mm × 270 mm × 368 mm
General description
Boro 3.3. DIN 12491. Precisely ground flat flanges. Base ring surface studded to improve stability and help prevent cracks. Interchangeable lid. Desiccators with sockets are supplied with 24/29 socket, and interchangeable Boro 3.3 stopcock.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)