Key Documents
Z742416
PTFE-faced silicone septa for Q-Tube and QianCap
septa 12 mL, max. pressure 180 psi
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
material
PTFE face
white silicone backing
manufacturer/tradename
Q Labtech 100-000825-001
parameter
180 psi max. pressure
General description
This disposable septa are made of tan PTFE face and white silicone backing. It fits on Q-Tube-12 mL with Adapter-180 psi. The tan PTFE liner has excellent chemical resistance and silicon backing has reseal ability. Using other septa may cause explosion.
Legal Information
Q-Tube is a trademark of Q Labtech LLC
QianCap is a trademark of Q Labtech LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)