Documents
BR470045
BRAND® cover glasses for microscope slides
L × W 18 mm × 18 mm, square
Synonym(s):
Cover glasses, Microscopy
About This Item
UNSPSC Code:
41122600
NACRES:
MA.01
material
borosilicate glass
square
manufacturer/tradename
BRAND 470045
L × W
18 mm × 18 mm
thickness
0.13-0.17 mm , No. 1
General description
Clear borosilicate glass, hydrolytic class 1, with excellent chemical resistance. Thickness No. 1 (0.13 to 0.17 mm). Refractive index ne = 1.52 ± 0.01, Abbe number ve = 56.5 ± 0.5. Full automatic production guarantees "ready-for-use" quality - clean, dust and grease free, distortion-free, flatness within ± 3 µm.
Features and Benefits
  • Ready-for-use
  • Made of hydrolytic class 1 glass with great chemical resistance
  • Clean: dust and grease free
  • Distortion-free
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG