Key Documents
BR4600101
BRAND® Dispensette® S Analog-adjustable bottle-top dispenser
volume range 0.1-1 mL, with recirculation valve
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
feature
incremental 0.02 mL
DE-M marking
with recirculation valve
manufacturer/tradename
BRAND 4600101
volume range
0.1-1 mL
fitting
(GL 24-25)
(GL 28/S 28)
GL 32
GL 38
S 40
General description
Dispensette S is suitable for an amazingly broad range of applications, particularly for many acids, saline solutions and organic solvents.
Features and Benefits
  • Extremely easy filling and dispensing
  • Simple priming by optimized flow channels
  • Rapid and safe volume adjustment with internal scalloped track
Other Notes
Analog-adjustable - Rapid and safe volume setting with new internal scalloped track. Simple calibration adjustment with a supplied tool.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
Visit the Document Library
Protocols
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.
Related Content
Chemical compatibility chart for all Dispensette® S and Dispensette® S Organic models.