Skip to Content
MilliporeSigma
All Photos(1)

Documents

BR38208

BRAND® volumetric flasks with 3 marks

capacity 50 mL, joint: 12/21, polypropylene stopper

Synonym(s):

BLAUBRAND®, DAkkS, Volumetric flasks

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121804
NACRES:
NB.01

material

borosilicate glass 3.3 flask
polypropylene stopper

Joint

joint ST/NS neck12/21
joint: 12/21

feature

class A DE-M marking, DIN EN ISO 1042. Calibrated to contain (TC, In). Incl. DAkkS-certificate.

packaging

pack of 1 ea

manufacturer/tradename

BRAND 38208

capacity

50 mL

Looking for similar products? Visit Product Comparison Guide

General description

Boro 3.3, DIN EN ISO 1042. Calibrated to contain (TC, In). Incl. DAkkS-certificate. The volumetric flask with 3 marks is used to check the functioning of a dispenser. The mark in the middle shows the nominal volume, the upper and lower marks show the error limits as specified in the table below. If the error limit is exceeded even with repeated measurements, the dispenser is out of specification. The DAkkS-calibrated volumetric flask does not replace the gravimetric test of the monitoring of measuring instruments according to ISO 8655.

Features and Benefits

  • BRAND® volumetric flasks with 3 marks are used for the accurate measurement of instruments for precision analyses.       
  • Calibrated to contain (TC, In).
  • The volumetric flask is used to check the functioning of a dispenser. The mark in the middle shows the nominal volume, the upper and lower marks show the error limits. If the error limit is exceeded even with repeated measurements, the dispenser is out of specification.
  • The DAkkS-calibrated volumetric flask does not replace the gravimetric test of the monitoring of measuring instruments according to ISO 8655.

Legal Information

BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service