Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BR36841

BRAND® volumetric flask BLAUBRAND® ETERNA

class A, DE-M marking, capacity 25 mL, borosilicate glass

Synonym(s):

BLAUBRAND®, ETERNA, Volumetric flasks

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121804
NACRES:
NB.43

material

borosilicate glass
polypropylene stopper

Joint

joint ST/NS neck10/19

feature

accuracy class A

packaging

pack of 2

manufacturer/tradename

BRAND 36841

capacity

25 mL

Looking for similar products? Visit Product Comparison Guide

General description

The BRAND® volumetric flask BLAUBRAND® ETERNA is constructed from high-quality Boro 3.3 glass and adheres to DIN EN ISO 1042 standards. It is calibrated to contain (TC, In), ensuring precise liquid measurement for various laboratory applications. Each flask is delivered with a batch number and includes one batch certificate per packing unit from the manufacturer, providing quality assurance and traceability.

Legal Information

BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service