Key Documents
BR13936
BRAND® spare burette tubes for compact burette
size 25 mL, SILBERBRAND, amber AR-Glass
Synonym(s):
Accessories, Burettes, Compact Burettes, SILBERBRAND
About This Item
UNSPSC Code:
41121808
NACRES:
NB.43
material
amber AR-Glass
packaging
pack of 1 ea
manufacturer/tradename
BRAND 13936
L
410 mm
size
25 mL
Related Categories
General description
The BRAND® spare burette tubes for compact burettes are crafted from high-quality Boro 3.3 clear glass, ensuring both durability and visibility for accurate liquid measurement. These spare tubes are specifically designed to be compatible with BLAUBRAND® and SILBERBRAND compact burettes, making them a reliable choice for maintaining your liquid handling systems.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)