Skip to Content
MilliporeSigma
All Photos(1)

Documents

BR115520

BRAND® test tube rack

PTFE, Holds 4 x 30 mm tubes

Synonym(s):

Accessories, Test Tube Racks, Tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122804
NACRES:
NB.35

material

PTFE

feature

Holds 4 x 30 mm tubes

packaging

pack of 1 ea

manufacturer/tradename

BRAND 115520

L × W × H

180 mm × 60 mm × 80 mm

Looking for similar products? Visit Product Comparison Guide

General description

The BRAND® test tube rack is constructed from high-quality PTFE, ensuring excellent chemical resistance for a variety of laboratory applications. This robust rack is designed to withstand operating temperatures ranging from -200 to 250 °C, making it suitable for both extreme cold and high-heat environments. Its durable design provides secure support for test tubes, preventing spills and breakage during experiments. The non-reactive nature of PTFE ensures that samples remain uncontaminated, enhancing the reliability of your laboratory work.

Features and Benefits

  • BRAND® test tube racks are sturdy with excellent mechanical strength.
  • Chemically resistant to most reagents.
  • Suitable for use in temperatures from -200 °C to 250 °C.

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service