Key Documents
BR114817
BRAND® PP graduated centrifuge tube, screw cap
volume 15 mL, without base, non-sterile
Synonym(s):
Centrifuge tubes, Tubes
About This Item
UNSPSC Code:
41121703
NACRES:
NB.11
material
polypropylene
screw top cap
translucent tube
sterility
non-sterile
feature
RCF 3,000 × g
without base
packaging
pack of 750
manufacturer/tradename
BRAND 114817
volume
15 mL
General description
The BRAND® PP Graduated Centrifuge Tube with Screw Cap is made from high-quality polypropylene (PP) and features a translucent design for easy visibility of contents. This tube is engineered to withstand relative centrifugal forces (RCF) of up to 3000, making it suitable for various laboratory applications. It comes graduated, allowing for precise measurements, and is available in both non-sterile and gamma-ray sterilized options in 15 ml (graduated up to 13 ml) or 50 ml capacities. The screw cap, made from polyethylene (PE) and equipped with a sealing cone, ensures a secure closure to prevent leaks. This centrifuge tube offers high clarity and reliability, making it an essential tool for researchers and technicians in the lab.
Features and Benefits
  • BRAND® graduated centrifuge tube is the ideal solution for all your centrifuge needs.
  • Made from highly transparent polypropylene (PP) for enhanced sample visibility.
  • Bio-inert PP offers optimal cell and protein recovery.
  • Includes a screw cap with a plug for preventing sample loss.
  • Tubes are tested to withstand RCF up to 3000.
  • γ-ray sterilized.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Articles
Find more about the analysis of 16 PFAS in salmon via QuEChERS extraction and LC-MS/MS characterization following FDA Method C-010.02.
Find more about the analysis of 16 PFAS in salmon via QuEChERS extraction and LC-MS/MS characterization following FDA Method C-010.02.
Find more about the analysis of 16 PFAS in salmon via QuEChERS extraction and LC-MS/MS characterization following FDA Method C-010.02.
Find more about the analysis of 16 PFAS in salmon via QuEChERS extraction and LC-MS/MS characterization following FDA Method C-010.02.
See All
Related Content
FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.
FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.