Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BR114715

BRAND® universal sample tube

diam. × L 16 mm × 100 mm

Synonym(s):

Test tubes, Tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.51

material

clear
colorless tube
polystyrene tube
round bottom

description

RCF max - 2000

sterility

non-sterile

feature

RCF 2,000 × g
graduations: no

packaging

pack of 3000 ea

manufacturer/tradename

BRAND 114715

diam. × L

16 mm × 100 mm

Looking for similar products? Visit Product Comparison Guide

General description

The BRAND® universal sample tube is constructed from transparent polystyrene (PS), providing clear visibility for monitoring samples. Its versatile design makes it suitable for a wide range of laboratory applications, from sample collection to storage.

Features and Benefits

Excellent sample visibility

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service