Key Documents
BMSBR5000STACK
Sigma-Aldrich
Everlast Stacking platform with flat mat with 3.5" clearance
About This Item
UNSPSC Code:
41103811
NACRES:
NB.73
manufacturer/tradename
Benchmark Scientific BR5000-STACK
compatibility
BMSB3D5000, BMSB3D5000E, BMSB3D5000EUK
General description
Optional Stacking Platform with Flat Mat workable clearance 3.5 inches and maximum clearance of 4.5 inches.
Legal Information
Everlast is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)