MilliporeSigma
All Photos(4)
Key Documents
BAF371250014
Spinfin® magnetic stirring bar
H × diam. 0.375 in. (9.5 mm) × 0.3125 in. (7.9 mm)
Synonym(s):
Spinfin® magnetic stirring bar
About This Item
UNSPSC Code:
41122800
NACRES:
NB.51
material
PTFE-coated
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F37125-0014
H × diam.
0.375 in. (9.5 mm) × 0.3125 in. (7.9 mm)
General description
Disks have fins on top for complete agitation.
Spinfin®magnetic stirring bars agitate liquids in round bottom vessels such as test tubes, cylinders and flasks, where other stirring bars cannot be used.
Features and Benefits
  • Maintains its position in a vessel and will not wobble
  • Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE® brand PTFE Coating – Superior Chemical Compatibility
  • Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification
  • Autoclave safe
Legal Information
Spinfin is a registered trademark of SP Scienceware, a division of SP
Teflon is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)