Key Documents
BAF201971515
SP Bel-Art® Safe-Lab Hollow PTFE Stoppers
for for 24/40 Tapered Joints
About This Item
UNSPSC Code:
41122800
NACRES:
NB.43
packaging
pack of 2 ea
manufacturer/tradename
Bel-Art F20197-1515
fitting
for for 24/40 Tapered Joints
General description
Use these hollow PTFE stoppers for boiling and distilling flasks with ground joints.
Features and Benefits
  • Easily removed from standard tapered glassware by turning the red extraction nut to apply a safe, gentle and even force against the mouth of the vessel
  • Helps eliminate accidents and breakage
  • Autoclave safe at 121ºC (250ºF)
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)