Key Documents
BAF196030000
SP Bel-Art® “T” Shaped Tubing Connectors
for ³⁄₁₆ in. Tubing
Synonym(s):
connectors, tubing, tubing coupler
About This Item
UNSPSC Code:
41122800
NACRES:
NB.43
packaging
pack of 12 ea
manufacturer/tradename
Bel-Art F19603-0000
I.D.
4.7 mm (0.1875 in.)
L × width
1.5 in. (38.1 mm) × 0.875 in. (22.2 mm)
General description
Connectors have multiple barbs per arm providing additional contact between the fitting and the tubing for extra gripping power.
Features and Benefits
  • Ensures a tight, secure connection between lengths of tubing having the same inner diameter (I.D.)
  • Chemical-resistant polypropylene for use with a wide range of liquids or gases
  • FDA-grade for direct contact with food, drugs, and cosmetics
  • Autoclavable
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP