Key Documents
BAF187510160
Scienceware® POXYGRID® test-tube rack
Holds 60 x 10-13 mm tubes, blue
Synonym(s):
tube holder
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
material
blue
epoxy (coated steel)
feature
Holds 60 x 10-13 mm tubes
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F18751-0160
L × W × H
17.5 cm × 11.4 cm × 6.4 cm
General description
Welded steel wire is coated with heat-cured, thermosetting epoxy to protect from rust and corrosion. The coating is unaffected by most organic solvents, weak acids and alkalis, and has excellent abrasion resistance and adhesion to steel substrate.
Temp. range: -370 to 250°F (-233 to 121°C). Has square holes, three decks, slotted-mesh bottom deck and reinforced edges.
Legal Information
Poxygrid is a registered trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)