Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D44804

Sigma-Aldrich

α,α′-Dibromo-p-xylene

97%

Synonym(s):

1,4-Bis(bromomethyl)benzene, p-Xylylene dibromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH2Br)2
CAS Number:
Molecular Weight:
263.96
Beilstein:
1100019
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

crystals

bp

245 °C (lit.)

mp

143-145 °C (lit.)

SMILES string

BrCc1ccc(CBr)cc1

InChI

1S/C8H8Br2/c9-5-7-1-2-8(6-10)4-3-7/h1-4H,5-6H2

InChI key

RBZMSGOBSOCYHR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

α,α′-Dibromo-p-xylene, also known as p-xylylene dibromide, is an aliphatic dihalide that serves as a versatile intermediate in organic reactions, including the synthesis of aromatic ionene polymers and dual-site phase-transfer catalysts.

Application

α,α′-Dibromo-p-xylene can be used as a building block to synthesize:      
  • Phthalazinone derivatives, which are reported as potent inhibitors of phosphodiesterase 4.    
  • N-Phenylated polyamine via polycondensation reaction with aniline and α, α′-dianilino-p-xylene.     
  • Porous polymeric network (PPN) via a Friedel-Crafts alkylation reaction with triphenylamine using FeCl3 as an oxidizing agent.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service