Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B65209

Sigma-Aldrich

4-Bromodiphenyl ether

99%

Synonym(s):

4-Bromophenyl phenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4OC6H5
CAS Number:
Molecular Weight:
249.10
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.607 (lit.)

bp

305 °C (lit.)

mp

18 °C (lit.)

density

1.423 g/mL at 25 °C (lit.)

SMILES string

Brc1ccc(Oc2ccccc2)cc1

InChI

1S/C12H9BrO/c13-10-6-8-12(9-7-10)14-11-4-2-1-3-5-11/h1-9H

InChI key

JDUYPUMQALQRCN-UHFFFAOYSA-N

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Man Cai et al.
Environmental science and pollution research international, 22(18), 14258-14268 (2015-05-15)
Populus species are very effective in remediation of contaminants. Polybrominated diphenyl ethers (PBDEs) are commonly used as flame retardants and are known to be persistent environmental pollutants. Numerous studies have shown that PBDEs are rising in human tissues and biota.
Sheng Wang et al.
Environmental science and pollution research international, 22(21), 16914-16926 (2015-06-26)
Commonly used flame retardants, such as polybrominated diphenyl ethers, are extremely persistent in the environment, causing serious environmental risks. Certain strains of bacteria are able to degrade several low brominated congeners of PBDEs aerobically. However, the aerobic degradation pathway is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service