Skip to Content
MilliporeSigma
All Photos(1)

Documents

252247

Sigma-Aldrich

2-Bromobiphenyl

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H4Br
CAS Number:
Molecular Weight:
233.10
Beilstein:
1863156
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

liquid

refractive index

n20/D 1.628 (lit.)

bp

297-298 °C (lit.)

mp

1.5-2 °C (lit.)

density

1.352 g/mL at 25 °C (lit.)

SMILES string

Brc1ccccc1-c2ccccc2

InChI

1S/C12H9Br/c13-12-9-5-4-8-11(12)10-6-2-1-3-7-10/h1-9H

InChI key

KTADSLDAUJLZGL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Bromobiphenyl has been employed:
  • as reference compound to investigate extraction efficiency of solid-phase microextraction fibers, based on methacrylic acid-trimethylolpropanetrimethacrylate copolymers
  • in synthesis of a novel trispirocyclic hydrocarbon having three 9-fluorene moieties around the core of truxene

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jingbin Zeng et al.
Analytica chimica acta, 648(2), 194-199 (2009-08-04)
The extraction performance of solid-phase microextraction fibers, based on methacrylic acid-trimethylolpropanetrimethacrylate (MAA/TRIM) copolymers, was investigated in aqueous solutions using triazines as target analytes and 2-bromobiphenyl as a reference compound. The results revealed that the combination of electrostatic forces and hydrophobicity
New 9-fluorene-type trispirocyclic compounds for thermally stable hole transport materials in OLEDs.
Kimura M, et al.
Journal of Materials Chemistry, 15(24), 2393-2398 (2005)
Michael Barclay et al.
Physical chemistry chemical physics : PCCP, 21(8), 4556-4567 (2019-02-12)
We present a combined theoretical and experimental study on the ionization and primary fragmentation channels of the mono-halogenated biphenyls; 2-chlorobiphenyl, 2-bromobiphenyl and 2-iodobiphenyl. The ionization energies (IEs) of the 2-halobiphenyls and the appearance energies (AEs) of the principal fragments are
Man Wang et al.
Journal of chromatography. A, 1412, 12-21 (2015-08-19)
A new magnetic porous carbon-doped graphitic carbon nitride nanocomposite and experimental strategies were environment-friendly designed for solid phase extraction of brominated flame retardants from water sample. The easily synthesized and low cost nanocomposite was characterized using techniques, including Fourier transform

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service