Documents
ACE729805
Ace addition funnel
capacity 125 mL
About This Item
UNSPSC Code:
41106514
NACRES:
NB.43
feature
0-3 pressure equalizing funnel
capacity
125 mL
General description
Angled position makes manipulation of stopcock easier than conventional style. Leak-proof with complete drainage, pressure-equalizing arm, precise needle-valve adjustment, and double-scale graduations.
Threaded stopcock (0 to 3 mm orifice for funnels up to 250 mL and 0 to 5 mm orifice for funnels 500 mL and larger) with PTFE plug permits smooth needle-valve adjustment down to 0.1 mL/min flow rate. Double PTFE-ring seals prevent exposure of backup O-ring to corrosive liquids.