A1401
2-Acetamidoacrylic acid
99%
Synonym(s):
N-Acetyldehydroalanine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH2=C(NHCOCH3)COOH
CAS Number:
Molecular Weight:
129.11
Beilstein:
1812032
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99%
form
powder
mp
185-186 °C (dec.) (lit.)
SMILES string
CC(=O)NC(=C)C(O)=O
InChI
1S/C5H7NO3/c1-3(5(8)9)6-4(2)7/h1H2,2H3,(H,6,7)(H,8,9)
InChI key
UFDFFEMHDKXMBG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Kaixin Fu et al.
Biosensors & bioelectronics, 143, 111636-111636 (2019-09-03)
Ketamine is one of the most widely abused drugs in the world and poses a serious threat to human health and social stability; therefore, the ability to accurately monitor the substance in real-time is necessary. However, several problems still exists
Shahaji R Gaikwad et al.
ChemPlusChem, 85(6), 1200-1209 (2020-06-26)
Insertion or coordination copolymerization of ethylene with di-substituted olefins is challenging and the choice of di-substituted mono-functional olefin versus di-substituted di-functional olefin (DDO) appears to be decisive. Here we show that DDO-inserted species are amenable to ethylene insertion and polymerization.
Hu-Chun Tao et al.
Nanoscale research letters, 12(1), 583-583 (2017-11-08)
Three kinds of functional monomers, 4-vinylpridine(4-VP), 2-(allylthio)nicotinic acid(ANA), and 2-Acetamidoacrylic acid(AAA), were used to synthetize palladium(II) ion-imprinted polymeric nanospheres (Pd(II) IIPs) via precipitation-polymerization method in order to study the effects of different functional monomers on the adsorption properties of ion-imprinted
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service