Skip to Content
MilliporeSigma
All Photos(1)

Documents

95889

Sigma-Aldrich

(S)-(−)-4-Methoxy-α-methylbenzylamine

≥98.0% (sum of enantiomers, GC)

Synonym(s):

(S)-(−)-1-(4-Methoxyphenyl)ethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13NO
CAS Number:
Molecular Weight:
151.21
Beilstein:
3196456
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (sum of enantiomers, GC)

form

liquid

optical activity

[α]20/D −32±2°, neat

optical purity

enantiomeric ratio: ≥97.5:2.5 (GC)

refractive index

n20/D 1.533

density

1.024 g/mL at 20 °C (lit.)

SMILES string

COc1ccc(cc1)[C@H](C)N

InChI

1S/C9H13NO/c1-7(10)8-3-5-9(11-2)6-4-8/h3-7H,10H2,1-2H3/t7-/m0/s1

InChI key

JTDGKQNNPKXKII-ZETCQYMHSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service