Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

938378

Sigma-Aldrich

BMB Crosslinker

≥95%

Synonym(s):

1,1′-(1,4-Butanediyl)bis[1H-pyrrole-2,5-dione], 1H-Pyrrole-2,5-dione, 1,1′-(1,4-butanediyl)bis-, Maleimide, N,N′-tetramethylenedi-, Tetramethylenedimaleimide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12N2O4
CAS Number:
Molecular Weight:
248.23
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥95%

form

powder or crystals

color

white to beige

mp

206 °C

storage temp.

2-8°C

SMILES string

O=C1C=CC(=O)N1CCCCN2C(=O)C=CC2=O

InChI

InChI=1S/C12H12N2O4/c15-9-3-4-10(16)13(9)7-1-2-8-14-11(17)5-6-12(14)18/h3-6H,1-2,7-8H2

InChI key

WXXSHAKLDCERGU-UHFFFAOYSA-N

Application

BMB crosslinker is a bismaleimide homobifunctional crosslinking reagent useful for conjugation to sulhydryl groups.

Features and Benefits

Useful for characterization of protein structures and interactions. A non cleavable 4-atom branched spacer arm assists in close proximity crosslinking.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Furan-maleimide thermoplast-thermoset merged polyimides via furan-maleimide Diels?Alder polymerization
Yogesh S Patel, et al.
International Journal of Plastics Technology, 17, 10-23 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service