Skip to Content
MilliporeSigma
All Photos(1)

Documents

936634

Sigma-Aldrich

1,6-BMH crosslinker

≥95%

Synonym(s):

1,1′-(1,6-Hexanediyl)bis[1H-pyrrole-2,5-dione] (ACI), 1,6-Bis(maleimido)hexane, 1H-Pyrrole-2,5-dione, 1,1′-(1,6-hexanediyl)bis- (9CI, ACI), N,N′-Hexamethylenebis[maleimide], Maleimide, N,N′-hexamethylenedi- (6CI, 7CI, 8CI)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16N2O4
CAS Number:
Molecular Weight:
276.29
MDL number:
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder or crystals

color

white to beige

mp

136-141 °C

storage temp.

2-8°C

SMILES string

O=C1C=CC(=O)N1CCCCCCN2C(=O)C=CC2=O

InChI

InChI=1S/C14H16N2O4/c17-11-5-6-12(18)15(11)9-3-1-2-4-10-16-13(19)7-8-14(16)20/h5-8H,1-4,9-10H2

Application

1, 6- BMH crosslinker is a bismaleimide homobifunctional crosslinking reagent useful for conjugation to sulhydryl groups.

Features and Benefits

Useful for characterization of protein structures and interactions. A non cleavable 6-atom brached spacer arm assists in close proximity crosslinking.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Furan-maleimide thermoplast-thermoset merged polyimides via furan-maleimide Diels?Alder polymerization
Patel S Y, et al
International Journal of Plastics Technology, 17, 10-23 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service