Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

936626

Sigma-Aldrich

Fmoc-Val-Cit-PAB-PNP

≥95%

Synonym(s):

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-valyl-N5-(aminocarbonyl)-N-[4-[[[(4-nitrophenoxy)carbonyl]oxy]methyl]phenyl]-L-ornithinamide (ACI), L-Ornithinamide, N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-valyl-N5-(aminocarbonyl)-N-[4-[[[(4-nitrophenoxy)carbonyl]oxy]methyl]phenyl]- (9CI, ACI)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H42N6O10
CAS Number:
Molecular Weight:
766.80
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder or crystals

color

Off White to Yellow to Beige

storage temp.

2-8°C

SMILES string

C(OC(N[C@H](C(N[C@H](C(NC1=CC=C(COC(OC2=CC=C(N(=O)=O)C=C2)=O)C=C1 )=O)CCCNC(N)=0)=O)C(C)C)=O)C3C=4C(C=5C3-CC-CC5)=CC-CC4

InChI

1S/C40H42N6O10/c1-24(2)35(45-39(50)54-23-33-31-10-5-3-8-29(31)30-9-4-6-11-32(30)33)37(48)44-34(12-7-21-42-38(41)49)36(47)43-26-15-13-25(14-16-26)22-55-40(51)56-28-19-17-27(18-20-28)46(52)53/h3-6,8-11,13-20,24,33-35H,7,12,21-23H2,1-2H3,(H,43,47)(H,44,48)(H,45,50)(H3,41,42,49)/t34-,35-/m0/s1

InChI key

USMYACISHVPTHK-PXLJZGITSA-N

Application

This is a cathepsin cleavable linker used in bioconjugation, specifically in the construction of antibody drug conjugates (ADC & Bioconjugation).

Features and Benefits

Fmoc-Val-Cit-PAB-PNP is a cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADC). The Val-Cit is specifically cleaved by cathepsin B. Fmoc-Val-Cit-PAB-PNP has superior plasma stability comparable to that of non-cleavable linkers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zixuan Ye et al.
International journal of nanomedicine, 16, 2443-2459 (2021-04-06)
Specific modifications to carriers to achieve targeted delivery of chemotherapeutics into malignant tissues are a critical point for efficient diagnosis and therapy. In this case, bovine serum albumin (BSA) was conjugated with cetuximab-valine-citrulline (vc)-doxorubicin (DOX) to target epidermal growth factor
Qinhuai Lai et al.
European journal of medicinal chemistry, 199, 112364-112364 (2020-05-14)
Cryptophycin-52 (CR52), a tubulin inhibitor, exhibits promising antitumor activity in vitro (picomolar level) and in mouse xenograft models. However, the narrow therapeutic window in clinical trials limits its further development. Antibody-drug conjugate (ADC), formed by coupling cytotoxic compound (payload) to an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service