Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

936375

Sigma-Aldrich

DBCO Acid

≥95%

Synonym(s):

11,12-Didehydro-γ-oxodibenz[b,f]azocine-5(6H)-butanoic acid (ACI), Dibenz[b,f]azocine-5(6H)-butanoic acid, 11,12-didehydro-γ-oxo- (ACI)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H15NO3
CAS Number:
Molecular Weight:
305.33
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder or crystals

color

white to off-white

mp

163-164 °C

storage temp.

−20°C

SMILES string

OC(CCC(N1CC2=CC=CC=C2C#CC3=CC=CC=C13)=O)=O

InChI

InChI=1S/C19H15NO3/c21-18(11-12-19(22)23)20-13-16-7-2-1-5-14(16)9-10-15-6-3-4-8-17(15)20/h1-8H,11-13H2,(H,22,23)

Application

DBCO Acid is a bifunctional building block comprising of an acid terminus useful for reaction with amine containing linkers in presence of activators such as EDC or HATU and a dibenzocyclooctyne (DBCO) terminus useful for copper-free click chemistry.

Features and Benefits

DBCO compounds react with azides without need of Cu(I) catalyst due to the strained nature of alkyne resulting in formation of fast and stable triazole linkage.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Copper-free click chemistry in living animals
Chang P V, et al.
Proceedings of the National Academy of Sciences of the USA, 107, 1821-1826 (2010)
A stepwise huisgen cycloaddition process: copper(I)-catalyzed regioselective "ligation" of azides and terminal alkynes
Rostovtsev V V, et al.
Angewandte Chemie (International ed. in English), 41, 2596-2599 (2002)
Pamela V Chang et al.
Proceedings of the National Academy of Sciences of the United States of America, 107(5), 1821-1826 (2010-01-19)
Chemical reactions that enable selective biomolecule labeling in living organisms offer a means to probe biological processes in vivo. Very few reactions possess the requisite bioorthogonality, and, among these, only the Staudinger ligation between azides and triarylphosphines has been employed
A stepwise huisgen cycloaddition process: copper(I)-catalyzed regioselective "ligation" of azides and terminal alkynes.
Vsevolod V Rostovtsev et al.
Angewandte Chemie (International ed. in English), 41(14), 2596-2599 (2002-08-31)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service