Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

932310

Sigma-Aldrich

Lithium fluoride

Synonym(s):

LiF, Fluorolithium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiF
CAS Number:
Molecular Weight:
25.94
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.23
Assay:
≥99%
solubility:
H2O: 2.9 g/L

Assay

≥99%

Quality Level

bp

1681 °C

mp

845 °C (lit.)

solubility

H2O: 2.9 g/L

density

2.64 g/mL at 25 °C (lit.)

Orbital energy

HOMO 14 eV 
LUMO 1.0 eV 

SMILES string

[Li+].[F-]

InChI

1S/FH.Li/h1H;/q;+1/p-1

InChI key

PQXKHYXIUOZZFA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

LiF can be used in thermoluminescent; perovskite light-emitting diodes; rechargeable batteries and MXenes applications. Lithium fluoride crystals are transparent to ultraviolet (UV) light and are used in UV optics. Lithium fluoride is used in the main route of fabrication of Mxenes by exfoliating MAX phases.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recent progress in LiF materials for safe lithium metal anode of rechargeable batteries: Is LiF the key to commercializing Li metal batteries?
Ko J, et al.
Ceramics International, 45(1), 30-49 (2019)
Mingfu He et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(1), 73-79 (2019-12-19)
Lithium is the most attractive anode material for high-energy density rechargeable batteries, but its cycling is plagued by morphological irreversibility and dendrite growth that arise in part from its heterogeneous "native" solid electrolyte interphase (SEI). Enriching the SEI with lithium
Bing Zhou et al.
ACS applied materials & interfaces, 12(4), 4895-4905 (2020-01-04)
Flexible, lightweight, robust, and multifunctional characteristics are greatly desirable for next-generation wearable electromagnetic interference (EMI) shielding materials. In this work, an alternating multilayered structure with robust polymer frame layers and directly contacted conducting layers was designed to prepare high-performance EMI
Xiaolei Yang et al.
Nature communications, 9(1), 570-570 (2018-02-10)
Perovskite light-emitting diodes (LEDs) are attracting great attention due to their efficient and narrow emission. Quasi-two-dimensional perovskites with Ruddlesden-Popper-type layered structures can enlarge exciton binding energy and confine charge carriers and are considered good candidate materials for efficient LEDs. However
Xiulin Fan et al.
Science advances, 4(12), eaau9245-eaau9245 (2018-12-28)
Solid-state electrolytes (SSEs) are receiving great interest because their high mechanical strength and transference number could potentially suppress Li dendrites and their high electrochemical stability allows the use of high-voltage cathodes, which enhances the energy density and safety of batteries.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service