Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

26158-U

Hamilton® plunger assemblies with tips for series 1000, 1700, and 1800

for use with Hamilton 1750 (LT, LTN, RN, or TLL), pkg of 1 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001
NACRES:
NB.03

material

PTFE plunger tip

size

500 μL

feature

1750 LT, LTN, RN, or TLL

packaging

pkg of 1 ea

manufacturer/tradename

Hamilton 1169-01

compatibility

for use with Hamilton 1750 (LT, LTN, RN, or TLL)

Looking for similar products? Visit Product Comparison Guide

General description

For field repair of Hamilton Series 1000, 1700 and 1800 syringes.

Legal Information

Hamilton is a registered trademark of Hamilton Co.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service