Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

932248

Sigma-Aldrich

Poly[(9,9-dioctyl-2,7-divinylenefluorenylene)-alt-{2-methoxy-5-(2-ethylhexyloxy)-1,4-phenylene}]

Synonym(s):

PFOPV

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C48H66O2)n
CAS Number:
UNSPSC Code:
12352207
NACRES:
NA.23

description

UV: 480 nm (in THF)

PL: 539 nm (in THF)

Eg = 2.3 eV

Quality Level

mol wt

Mw ≥20,000 Da by GPC

energy gap

2.3 eV (Eg (HOMO-LUMO gap))

solubility

THF: soluble
chloroform: soluble (or chlorobenzene)
toluene: soluble

λmax

480 nm±5 nm in THF

fluorescence

λem 508 nm±10 nm in THF
λem 539 nm±10 nm in THF

Orbital energy

HOMO 4.4 eV 
LUMO 2.1 eV 

Application

Polymer Emitting Layers for Solution-Processed OLEDs; green emission.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lasing and Transport Properties of Poly[(9,9-dioctyl-2,7-divinylenefluorenylene)-alt-co-(2-methoxy-5-(2-ethylhexyloxy)-1,4-phenylene)] (POFP) for the Application of Diode-Pumped Organic Solid Lasers
Tang Z, et al.
Nanoscale Research Letters, 12(1), 602-602 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service