Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

932000

Sigma-Aldrich

PSCbCz

Synonym(s):

NPI_emissivelayer, Poly{6-(9H-Carbazol-9-yl)-9-(4-vinylbenzyl)-9H-pyrido-[2,3-b]indole}

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C32H23N3)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

mol wt

≥9000 by GPC

Quality Level

energy gap

0.15 eV (delta EST)
3.38 eV (HOMO-LUMO gap)

solubility

chlorobenzene: soluble

fluorescence

λem 414 nm±10 nm in chlorobenzene

Orbital energy

HOMO -5.65 eV 
LUMO -2.28 eV 

Application

PSCbCz is a carbazole-based polymeric host materials for realizing high-efficiency solution-processed polymer host emitting layer for phosphorescent green TADF OLED, with external quantum efficiencies (EQE) of 17.48%.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinhyo Hwang et al.
ACS applied materials & interfaces, 12(7), 8485-8494 (2020-01-29)
Recently, various host materials have been developed for solution-processed thermally activated delayed fluorescent organic light-emitting diodes (TADF-OLEDs). Compared with small-molecule hosts, polymeric hosts are advantageous for inducing a uniform distribution and segregation of dopant molecules in the emissive layer without

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service