Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

932442

Sigma-Aldrich

4,4′-Bis(carbazol-9-yl)biphenyl

≥99% (HPLC)

Synonym(s):

4,4′-Bis(N-carbazolyl)-1,1′-biphenyl, 4,4′-Bis(9-carbazolyl)-1,1′-biphenyl, 4,4-N,N′-Dicarbazole-1,1′-biphenyl, CBP, DCBP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H24N2
CAS Number:
Molecular Weight:
484.59
MDL number:
UNSPSC Code:
12352005
NACRES:
NA.23
Assay:
≥99% (HPLC)
grade:
sublimed grade
solubility:
THF: soluble
chloroform: soluble
dichloromethane: soluble

grade

sublimed grade

Quality Level

description

µh ≈ 2.0 x 10-3cm2V-1S-1

Assay

≥99% (HPLC)

loss

0.5% TGA, >320 ºC (weight loss)

mp

281-285 °C

solubility

THF: soluble
chloroform: soluble
dichloromethane: soluble

fluorescence

λex 369 nm in THF

Orbital energy

HOMO 6.0 eV 
LUMO 2.9 eV 

λ

in THF

UV absorption

λ: 292 nm±5 nm Amax
λ: 318 nm±5 nm Amax

SMILES string

c1ccc2c(c1)n(-c3ccc(cc3)-c4ccc(cc4)-n5c6ccccc6c7ccccc57)c8ccccc28

InChI

1S/C36H24N2/c1-5-13-33-29(9-1)30-10-2-6-14-34(30)37(33)27-21-17-25(18-22-27)26-19-23-28(24-20-26)38-35-15-7-3-11-31(35)32-12-4-8-16-36(32)38/h1-24H

InChI key

VFUDMQLBKNMONU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4,4′-Bis(carbazol-9-yl)biphenyl can be used as a as a hole transport material or as part of the active layer inorganic photovoltaics (OPVs) devices. Due to its semiconducting properties, it can be used as a charge transport material in Organic Field-Effect Transistors(OFETs).It can be utilized as a host material in the emissive layer of organic light-emitting diodes (OLEDs) devices. It provides a suitable matrix for efficient energy transfer from dopant molecules, leading to enhanced electroluminescence and color purity in OLEDs.
4,4′-Bis(carbazol-9-yl)biphenyl, also known as CBP is used as solution-processable Phosphorescent Host in organic electronics such as OLED devices.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weixuan Zeng et al.
Advanced materials (Deerfield Beach, Fla.), 30(5) (2017-12-09)
The combination of rigid acridine donor and 1,8-naphthalimide acceptor has afforded two orange-red emitters of NAI-DMAC and NAI-DPAC with high rigidity in molecular structure and strongly pretwisted charge transfer state. Endowed with high photoluminescence quantum yields (ΦPL ), distinct thermally
Jia-Xiong Chen et al.
Angewandte Chemie (International ed. in English), 58(41), 14660-14665 (2019-07-18)
Developing red thermally activated delayed fluorescence (TADF) emitters, attainable for both high-efficient red organic light-emitting diodes (OLEDs) and non-doped deep red/near-infrared (NIR) OLEDs, is challenging. Now, two red emitters, BPPZ-PXZ and mDPBPZ-PXZ, with twisted donor-acceptor structures were designed and synthesized
Efficient blue organic light-emitting diodes employing thermally activated delayed fluorescence
Zhang Qisheng, et al.
Nature Photonics, 8, 326?332-326?332 (2014)
Ling Yu et al.
Chemical science, 9(5), 1385-1391 (2018-04-21)
Simultaneously achieving high efficiency and low efficiency roll-off remains a big challenge for OLEDs based on thermally activated delayed fluorescence (TADF) emitters. To address this issue, we elaborately designed and synthesized a series of new emitters with both TADF and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service