Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

929425

Sigma-Aldrich

Memantine-(dimethyl-d6) hydrochloride

≥97 atom % D, ≥95% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12D6H16ClN
Molecular Weight:
221.80
UNSPSC Code:
12352114
NACRES:
NA.74

isotopic purity

≥97 atom % D

Quality Level

Assay

≥95% (CP)

technique(s)

mass spectrometry (MS): suitable

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service